[2-Bromo-4-(difluoromethyl)-6-methoxy-phenyl] acetate

Lieferant: Apollo Scientific

PC520303-1G
APOSPC520303-1GEA 2280 EUR
APOSPC520303-1G
[2-Bromo-4-(difluoromethyl)-6-methoxy-phenyl] acetate
[2-Bromo-4-(difluoromethyl)-6-methoxy-phenyl] acetate
Formel: C₁₀H₉BrF₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD30496622
CAS-Nummer: 2149589-71-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR