1,3-Dibenzyloxy-5-(difluoromethyl)benzene

Lieferant: Apollo Scientific

PC520305-1G
APOSPC520305-1GEA 797 EUR
APOSPC520305-1G
1,3-Dibenzyloxy-5-(difluoromethyl)benzene
1,3-Dibenzyloxy-5-(difluoromethyl)benzene
Formel: C₂₁H₁₈F₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD30496623
CAS-Nummer: 2149601-37-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR