[3-(Chloromethyl)-5-(difluoromethyl)-4-hydroxy-phenyl]azinate

Lieferant: Apollo Scientific

PC520306-1G
APOSPC520306-1GEA 1610 EUR
APOSPC520306-1G
[3-(Chloromethyl)-5-(difluoromethyl)-4-hydroxy-phenyl]azinate
[3-(Chloromethyl)-5-(difluoromethyl)-4-hydroxy-phenyl]azinate
Formel: C₈H₆ClF₂NO₃
Molecular Weight: 237,588 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496625
CAS-Nummer: 2092612-17-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR