2-(Difluoromethyl)-4-iodo-phenol
Lieferant: Apollo Scientific
PC520307-1G
APOSPC520307-1GEA
1140
EUR
APOSPC520307-1G
2-(Difluoromethyl)-4-iodo-phenol
2-(Difluoromethyl)-4-iodo-phenol
Formel:
C₇H₅F₂IO Molecular Weight: 270,015 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767776 CAS-Nummer: 2092431-65-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...