5-(Difluoromethyl)-6-methoxy-1,3-benzodioxole
Lieferant: Apollo Scientific
PC520308-1G
APOSPC520308-1GEA
797
EUR
APOSPC520308-1G
5-(Difluoromethyl)-6-methoxy-1,3-benzodioxole
5-(Difluoromethyl)-6-methoxy-1,3-benzodioxole
Formel:
C₉H₈F₂O₃ Molecular Weight: 202,155 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767777 CAS-Nummer: 2149598-37-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...