1-(Difluoromethyl)-2-methyl-benzene

Lieferant: Apollo Scientific

PC520309-1G
APOSPC520309-1GEA 627 EUR
APOSPC520309-1G
1-(Difluoromethyl)-2-methyl-benzene
1-(Difluoromethyl)-2-methyl-benzene
Formel: C₈H₈F₂
Molecular Weight: 142,146 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767778
CAS-Nummer: 1222556-60-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR