1,3-bis(difluoromethyl)benzene
Lieferant: Apollo Scientific
PC520311-1G
APOSPC520311-1GEA
627
EUR
APOSPC520311-1G
1,3-bis(difluoromethyl)benzene
1,3-bis(difluoromethyl)benzene
Formel:
C₈H₆F₄ Molecular Weight: 178,127 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767780 CAS-Nummer: 936330-10-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...