tert-Butyl N-[2-(difluoromethyl)phenyl]carbamate

Lieferant: Apollo Scientific

PC520312-1G
APOSPC520312-1GEA 956 EUR
APOSPC520312-1G
tert-Butyl N-[2-(difluoromethyl)phenyl]carbamate
tert-Butyl N-[2-(difluoromethyl)phenyl]carbamate
Formel: C₁₂H₁₅F₂NO₂
Molecular Weight: 243,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767782
CAS-Nummer: 2002442-03-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR