Methyl 5-(difluoromethyl)-2-hydroxy-benzoate

Lieferant: Apollo Scientific

PC520316-1G
APOSPC520316-1GEA 1200 EUR
APOSPC520316-1G
Methyl 5-(difluoromethyl)-2-hydroxy-benzoate
Methyl 5-(difluoromethyl)-2-hydroxy-benzoate
Formel: C₉H₈F₂O₃
Molecular Weight: 202,155 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767784
CAS-Nummer: 2091158-31-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR