Methyl 3-bromo-5-(difluoromethyl)-2-hydroxy-benzoate

Lieferant: Apollo Scientific

PC520317-1G
APOSPC520317-1GEA 2280 EUR
APOSPC520317-1G
Methyl 3-bromo-5-(difluoromethyl)-2-hydroxy-benzoate
Methyl 3-bromo-5-(difluoromethyl)-2-hydroxy-benzoate
Formel: C₉H₇BrF₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD30496629
CAS-Nummer: 2091144-56-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR