2-(Difluoromethyl)-3-fluoro-4-methoxy-phenol

Lieferant: Apollo Scientific

PC520323-1G
APOSPC520323-1GEA 2270 EUR
APOSPC520323-1G
2-(Difluoromethyl)-3-fluoro-4-methoxy-phenol
2-(Difluoromethyl)-3-fluoro-4-methoxy-phenol
Formel: C₈H₇F₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD30496638
CAS-Nummer: 2090540-48-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR