4-benzyloxy-1-(difluoromethyl)-2-fluoro-benzene

Lieferant: Apollo Scientific

PC520324-1G
APOSPC520324-1GEA 2280 EUR
APOSPC520324-1G
4-benzyloxy-1-(difluoromethyl)-2-fluoro-benzene
4-benzyloxy-1-(difluoromethyl)-2-fluoro-benzene
Formel: C₁₄H₁₁F₃O
Molecular Weight: 252,232 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496639
CAS-Nummer: 2149601-32-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR