[4-(Difluoromethyl)-3-fluoro-phenyl]azinate

Lieferant: Apollo Scientific

PC520326-1G
APOSPC520326-1GEA 1200 EUR
APOSPC520326-1G
[4-(Difluoromethyl)-3-fluoro-phenyl]azinate
[4-(Difluoromethyl)-3-fluoro-phenyl]azinate
Formel: C₇H₄F₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD14698656
CAS-Nummer: 89641-03-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR