1-(4-Chlorophenoxy)-2-(difluoromethyl)-3-fluoro-benzene

Lieferant: Apollo Scientific

PC520329-1G
APOSPC520329-1GEA 1320 EUR
APOSPC520329-1G
1-(4-Chlorophenoxy)-2-(difluoromethyl)-3-fluoro-benzene
1-(4-Chlorophenoxy)-2-(difluoromethyl)-3-fluoro-benzene
Formel: C₁₃H₈ClF₃O
Molecular Weight: 272,65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496642
CAS-Nummer: 2149602-13-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR