5-Chloro-2-(difluoromethyl)-3-fluoro-phenol

Lieferant: Apollo Scientific

PC520333-1G
APOSPC520333-1GEA 2740 EUR
APOSPC520333-1G
5-Chloro-2-(difluoromethyl)-3-fluoro-phenol
5-Chloro-2-(difluoromethyl)-3-fluoro-phenol
Formel: C₇H₄ClF₃O
Molecular Weight: 196,554 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496645
CAS-Nummer: 2092102-01-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR