2-(Difluoromethyl)-3-fluoro-aniline

Lieferant: Apollo Scientific

PC520337-1G
APOSPC520337-1GEA 2740 EUR
APOSPC520337-1G
2-(Difluoromethyl)-3-fluoro-aniline
2-(Difluoromethyl)-3-fluoro-aniline
Formel: C₇H₆F₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD30496649
CAS-Nummer: 2091605-37-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR