[3-(Difluoromethyl)-2-fluoro-phenyl]azinate 97%

Lieferant: Apollo Scientific

PC520341-1G
APOSPC520341-1GEA 951 EUR
APOSPC520341-1G
[3-(Difluoromethyl)-2-fluoro-phenyl]azinate 97%
[3-(Difluoromethyl)-2-fluoro-phenyl]azinate
Formel: C₇H₄F₃NO₂
Molecular Weight: 191,107 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14698655
CAS-Nummer: 1214383-50-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR