4-Chloro-2-(difluoromethyl)-3-fluoro-phenol

Lieferant: Apollo Scientific

PC520342-1G
APOSPC520342-1GEA 2280 EUR
APOSPC520342-1G
4-Chloro-2-(difluoromethyl)-3-fluoro-phenol
4-Chloro-2-(difluoromethyl)-3-fluoro-phenol
Formel: C₇H₄ClF₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD30496651
CAS-Nummer: 2092380-93-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR