1-Chloro-4-(difluoromethyl)-5-fluoro-2-methoxy-benzene
Lieferant: Apollo Scientific
PC520344-1G
APOSPC520344-1GEA
1830
EUR
APOSPC520344-1G
1-Chloro-4-(difluoromethyl)-5-fluoro-2-methoxy-benzene
1-Chloro-4-(difluoromethyl)-5-fluoro-2-methoxy-benzene
Formel:
C₈H₆ClF₃O Molecular Weight: 210,581 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29767790 CAS-Nummer: 2149589-82-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...