2-(Difluoromethyl)-3-fluoro-6-nitro-phenol

Lieferant: Apollo Scientific

PC520345-1G
APOSPC520345-1GEA 2740 EUR
APOSPC520345-1G
2-(Difluoromethyl)-3-fluoro-6-nitro-phenol
2-(Difluoromethyl)-3-fluoro-6-nitro-phenol
Formel: C₇H₄F₃NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD30496652
CAS-Nummer: 2090420-72-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR