4-(Difluoromethyl)-5-fluoro-2-nitro-phenol

Lieferant: Apollo Scientific

PC520346-1G
APOSPC520346-1GEA 2740 EUR
APOSPC520346-1G
4-(Difluoromethyl)-5-fluoro-2-nitro-phenol
4-(Difluoromethyl)-5-fluoro-2-nitro-phenol
Formel: C₇H₄F₃NO₃
Molecular Weight: 207,107 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496653
CAS-Nummer: 2092102-06-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR