5-Chloro-2-(difluoromethyl)-1-fluoro-3-methoxy-benzene

Lieferant: Apollo Scientific

PC520347-1G
APOSPC520347-1GEA 2000 EUR
APOSPC520347-1G
5-Chloro-2-(difluoromethyl)-1-fluoro-3-methoxy-benzene
5-Chloro-2-(difluoromethyl)-1-fluoro-3-methoxy-benzene
Formel: C₈H₆ClF₃O
Molecular Weight: 210,581 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496654
CAS-Nummer: 2149590-26-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR