4-(Difluoromethyl)-1,5-dimethoxy-naphthalene

Lieferant: Apollo Scientific

PC520350-1G
APOSPC520350-1GEA 968 EUR
APOSPC520350-1G
4-(Difluoromethyl)-1,5-dimethoxy-naphthalene
4-(Difluoromethyl)-1,5-dimethoxy-naphthalene
Formel: C₁₃H₁₂F₂O₂
Molecular Weight: 238.23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29767792
CAS-Nummer: 2149597-07-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR