1-(2-Trifluoromethoxyphenyl) cyclopropanamine hydrochloride

Lieferant: Apollo Scientific

PC520351-1G
APOSPC520351-1GEA 1550 EUR
APOSPC520351-1G
1-(2-Trifluoromethoxyphenyl) cyclopropanamine hydrochloride
1-(2-Trifluoromethoxyphenyl) cyclopropanamine hydrochloride
Formel: C₁₀H₁₀F₃NO·ClH
Molecular Weight: 253,649 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496656
CAS-Nummer: 2149591-22-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR