1-(6-Fluoro-2-pyridyl)cyclopropanamine hydrochloride

Lieferant: Apollo Scientific

PC520352-1G
APOSPC520352-1GEA 2280 EUR
APOSPC520352-1G
1-(6-Fluoro-2-pyridyl)cyclopropanamine hydrochloride
1-(6-Fluoro-2-pyridyl)cyclopropanamine hydrochloride
Formel: C₈H₉FN₂·ClH
Molecular Weight: 188,63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496657
CAS-Nummer: 2149601-88-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR