1-(2-Fluoro-3-pyridyl)cyclopropanamine hydrochloride

Lieferant: Apollo Scientific

PC520354-1G
APOSPC520354-1GEA 2280 EUR
APOSPC520354-1G
1-(2-Fluoro-3-pyridyl)cyclopropanamine hydrochloride
1-(2-Fluoro-3-pyridyl)cyclopropanamine hydrochloride
Formel: C₈H₉FN₂·ClH
Molecular Weight: 188,63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496659
CAS-Nummer: 2149598-00-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR