1-(6-Fluoro-3-pyridyl)cyclopropanamine hydrochloride
Lieferant: Apollo Scientific
PC520357-1G
APOSPC520357-1GEA
2280
EUR
APOSPC520357-1G
1-(6-Fluoro-3-pyridyl)cyclopropanamine hydrochloride
1-(6-Fluoro-3-pyridyl)cyclopropanamine hydrochloride
Formel:
C₈H₉FN₂·ClH Molecular Weight: 188,63 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30496662 CAS-Nummer: 2149598-02-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...