1-(2-Fluoro-4-pyridyl)cyclopropanamine hydrochloride

Lieferant: Apollo Scientific

PC520359-1G
APOSPC520359-1GEA 2280 EUR
APOSPC520359-1G
1-(2-Fluoro-4-pyridyl)cyclopropanamine hydrochloride
1-(2-Fluoro-4-pyridyl)cyclopropanamine hydrochloride
Formel: C₈H₉FN₂·ClH
Molecular Weight: 188.63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496664
CAS-Nummer: 2149589-65-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR