1-[6-(Trifluoromethyl)-2-pyridyl]cyclopropanamine hydrochloride
Lieferant: Apollo Scientific
PC520360-1G
APOSPC520360-1GEA
2280
EUR
APOSPC520360-1G
1-[6-(Trifluoromethyl)-2-pyridyl]cyclopropanamine hydrochloride
1-[6-(Trifluoromethyl)-2-pyridyl]cyclopropanamine hydrochloride
Formel:
C₉H₉F₃N₂·ClH Molecular Weight: 238,637 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30496665 CAS-Nummer: 1384265-58-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...