1-[4-(Trifluoromethyl)-2-pyridyl]cyclopropanamine hydrochloride

Lieferant: Apollo Scientific

PC520362-1G
APOSPC520362-1GEA 2280 EUR
APOSPC520362-1G
1-[4-(Trifluoromethyl)-2-pyridyl]cyclopropanamine hydrochloride
1-[4-(Trifluoromethyl)-2-pyridyl]cyclopropanamine hydrochloride
Formel: C₉H₉F₃N₂·ClH
Molecular Weight: 238,637 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD30496667
CAS-Nummer: 1384264-67-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR