1-(5-Fluoro-3-pyridyl)cyclopropanecarbonitrile

Lieferant: Apollo Scientific

PC520370-1G
APOSPC520370-1GEA 2280 EUR
APOSPC520370-1G
1-(5-Fluoro-3-pyridyl)cyclopropanecarbonitrile
1-(5-Fluoro-3-pyridyl)cyclopropanecarbonitrile
Formel: C₉H₇FN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD23134891
CAS-Nummer: 1427010-14-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR