1-(6-Fluoro-3-pyridyl)cyclopropanecarbonitrile
Lieferant: Apollo Scientific
PC520371-1G
APOSPC520371-1GEA
2280
EUR
APOSPC520371-1G
1-(6-Fluoro-3-pyridyl)cyclopropanecarbonitrile
1-(6-Fluoro-3-pyridyl)cyclopropanecarbonitrile
Formel:
C₉H₇FN₂ Molecular Weight: 162,164 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23134892 CAS-Nummer: 1427011-98-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...