1-[2-(Trifluoromethyl)-4-pyridyl]cyclopropanecarbonitrile
Lieferant: Apollo Scientific
PC520372-1G
APOSPC520372-1GEA
3420
EUR
APOSPC520372-1G
1-[2-(Trifluoromethyl)-4-pyridyl]cyclopropanecarbonitrile
1-[2-(Trifluoromethyl)-4-pyridyl]cyclopropanecarbonitrile
Formel:
C₁₀H₇F₃N₂ Molecular Weight: 212,171 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23134895 CAS-Nummer: 1427024-70-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...