1-[2-(Trifluoromethyl)-3-pyridyl]cyclopropanecarbonitrile

Lieferant: Apollo Scientific

PC520373-1G
APOSPC520373-1GEA 3420 EUR
APOSPC520373-1G
1-[2-(Trifluoromethyl)-3-pyridyl]cyclopropanecarbonitrile
1-[2-(Trifluoromethyl)-3-pyridyl]cyclopropanecarbonitrile
Formel: C₁₀H₇F₃N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD23134897
CAS-Nummer: 1427012-75-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR