1-[4-(Trifluoromethyl)-3-pyridyl]cyclopropanecarbonitrile
Lieferant: Apollo Scientific
PC520374-1G
APOSPC520374-1GEA
3420
EUR
APOSPC520374-1G
1-[4-(Trifluoromethyl)-3-pyridyl]cyclopropanecarbonitrile
1-[4-(Trifluoromethyl)-3-pyridyl]cyclopropanecarbonitrile
Formel:
C₁₀H₇F₃N₂ Molecular Weight: 212,171 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23134898 CAS-Nummer: 1427014-29-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...