1-[3-(Trifluoromethyl)-2-pyridyl]cyclopropanecarbonitrile

Lieferant: Apollo Scientific

PC520377-1G
APOSPC520377-1GEA 3420 EUR
APOSPC520377-1G
1-[3-(Trifluoromethyl)-2-pyridyl]cyclopropanecarbonitrile
1-[3-(Trifluoromethyl)-2-pyridyl]cyclopropanecarbonitrile
Formel: C₁₀H₇F₃N₂
Molecular Weight: 212,171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23134901
CAS-Nummer: 1427012-98-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR