1-[5-(Trifluoromethyl)-2-pyridyl]cyclopropanecarbonitrile
Lieferant: Apollo Scientific
PC520378-1G
APOSPC520378-1GEA
3420
EUR
APOSPC520378-1G
1-[5-(Trifluoromethyl)-2-pyridyl]cyclopropanecarbonitrile
1-[5-(Trifluoromethyl)-2-pyridyl]cyclopropanecarbonitrile
Formel:
C₁₀H₇F₃N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD23134903 CAS-Nummer: 1427010-15-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...