1-(6-Fluoro-2-pyridyl)cyclopropanecarboxylic acid

Lieferant: Apollo Scientific

PC520380-1G
APOSPC520380-1GEA 1710 EUR
APOSPC520380-1G
1-(6-Fluoro-2-pyridyl)cyclopropanecarboxylic acid
1-(6-Fluoro-2-pyridyl)cyclopropanecarboxylic acid
Formel: C₉H₈FNO₂
Molecular Weight: 181,164 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23134921
CAS-Nummer: 1427011-62-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR