1-(4-Fluoro-3-pyridyl)cyclopropanecarboxylic acid

Lieferant: Apollo Scientific

PC520381-1G
APOSPC520381-1GEA 2740 EUR
APOSPC520381-1G
1-(4-Fluoro-3-pyridyl)cyclopropanecarboxylic acid
1-(4-Fluoro-3-pyridyl)cyclopropanecarboxylic acid
Formel: C₉H₈FNO₂
Molecular Weight: 181,164 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23134923
CAS-Nummer: 1427011-71-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR