1-(2-Fluoro-3-pyridyl)cyclopropanecarboxylic acid
Lieferant: Apollo Scientific
PC520382-1G
APOSPC520382-1GEA
2740
EUR
APOSPC520382-1G
1-(2-Fluoro-3-pyridyl)cyclopropanecarboxylic acid
1-(2-Fluoro-3-pyridyl)cyclopropanecarboxylic acid
Formel:
C₉H₈FNO₂ Molecular Weight: 181,164 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23134924 CAS-Nummer: 1427021-82-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...