1-(6-Fluoro-3-pyridyl)cyclopropanecarboxylic acid

Lieferant: Apollo Scientific

PC520383-1G
APOSPC520383-1GEA 2740 EUR
APOSPC520383-1G
1-(6-Fluoro-3-pyridyl)cyclopropanecarboxylic acid
1-(6-Fluoro-3-pyridyl)cyclopropanecarboxylic acid
Formel: C₉H₈FNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD23134925
CAS-Nummer: 1427010-46-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR