1-[2-(Trifluoromethyl)-4-pyridyl]cyclopropanecarboxylic acid

Lieferant: Apollo Scientific

PC520385-1G
APOSPC520385-1GEA 2740 EUR
APOSPC520385-1G
1-[2-(Trifluoromethyl)-4-pyridyl]cyclopropanecarboxylic acid
1-[2-(Trifluoromethyl)-4-pyridyl]cyclopropanecarboxylic acid
Formel: C₁₀H₈F₃NO₂
Molecular Weight: 231,171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23134928
CAS-Nummer: 1427013-07-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR