1-[3-(Trifluoromethyl)-4-pyridyl]cyclopropanecarboxylic acid

Lieferant: Apollo Scientific

PC520386-1G
APOSPC520386-1GEA 2740 EUR
APOSPC520386-1G
1-[3-(Trifluoromethyl)-4-pyridyl]cyclopropanecarboxylic acid
1-[3-(Trifluoromethyl)-4-pyridyl]cyclopropanecarboxylic acid
Formel: C₁₀H₈F₃NO₂
Molecular Weight: 231,171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23134929
CAS-Nummer: 1427024-73-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR