1-[2-(Trifluoromethyl)-3-pyridyl]cyclopropanecarboxylic acid
Lieferant: Apollo Scientific
PC520387-1G
APOSPC520387-1GEA
2740
EUR
APOSPC520387-1G
1-[2-(Trifluoromethyl)-3-pyridyl]cyclopropanecarboxylic acid
1-[2-(Trifluoromethyl)-3-pyridyl]cyclopropanecarboxylic acid
Formel:
C₁₀H₈F₃NO₂ Molecular Weight: 231,171 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23134930 CAS-Nummer: 1427012-25-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...