1-[4-(Trifluoromethyl)-3-pyridyl]cyclopropanecarboxylic acid

Lieferant: Apollo Scientific

PC520388-1G
APOSPC520388-1GEA 2740 EUR
APOSPC520388-1G
1-[4-(Trifluoromethyl)-3-pyridyl]cyclopropanecarboxylic acid
1-[4-(Trifluoromethyl)-3-pyridyl]cyclopropanecarboxylic acid
Formel: C₁₀H₈F₃NO₂
Molecular Weight: 231,171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23134931
CAS-Nummer: 1427024-29-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR