1-[3-(Trifluoromethyl)-2-pyridyl]cyclopropanecarboxylic acid

Lieferant: Apollo Scientific

PC520389-1G
APOSPC520389-1GEA 2740 EUR
APOSPC520389-1G
1-[3-(Trifluoromethyl)-2-pyridyl]cyclopropanecarboxylic acid
1-[3-(Trifluoromethyl)-2-pyridyl]cyclopropanecarboxylic acid
Formel: C₁₀H₈F₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD23134932
CAS-Nummer: 1427021-91-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR