N-[1-(4-Fluorophenyl)ethyl]prop-2-yn-1-amine hydrochloride

Lieferant: Apollo Scientific
Gefahr

PC520400-1G PC520400-250MG
APOSPC520400-1GEA 126 EUR
APOSPC520400-1G APOSPC520400-250MG
N-[1-(4-Fluorophenyl)ethyl]prop-2-yn-1-amine hydrochloride
N-[1-(4-Fluorophenyl)ethyl]prop-2-yn-1-amine hydrochloride
Formel: C₁₁H₁₂FN·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD31745722
CAS-Nummer: 2379918-45-1
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR