1-(2-Naphthoyl)-3,3,3-trifluoraceton 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4,4,4-Trifluoro-1-(naphthalen-2-yl)butane-1,3-dione, 4,4,4-Trifluoro-1-(2-naphthyl)-1,3-butanedione

PC5204M-100G PC5204M-25G PC5204M-500G
APOSPC5204M-100GEA 133 EUR
APOSPC5204M-100G APOSPC5204M-25G APOSPC5204M-500G
1-(2-Naphthoyl)-3,3,3-trifluoraceton 98%
1-(2-Naphthoyl)-3,3,3-trifluoraceton
Formel: C₁₄H₉F₃O₂
Molecular Weight: 266.22 g/mol
Schmelzpunkt: 70…76 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00054501
CAS-Nummer: 893-33-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR