4-Chlor-2-nitrobenzotrifluorid 95%

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-2-nitrobenzotrifluoride, 1-Chloro-3-nitro-4-(trifluoromethyl)benzene

PC520503-10G PC520503-1G PC520503-5G
APOSPC520503-10GEA 640 EUR
APOSPC520503-10G APOSPC520503-1G APOSPC520503-5G
4-Chlor-2-nitrobenzotrifluorid 95%
4-Chlor-2-nitrobenzotrifluorid
Formel: C₇H₃ClF₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00047693
CAS-Nummer: 25889-38-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR