Tris(2,2,3,3-tetrafluoropropyl) borate

Lieferant: Apollo Scientific
Achtung

Synonyme: Tris(2,2,3,3-tetrafluoropropoxy)borane

PC52051-5G
APOSPC52051-5GEA 607 EUR
APOSPC52051-5G
Tris(2,2,3,3-tetrafluoropropyl) borate
Tris(2,2,3,3-tetrafluoropropyl) borate
Formel: C₉H₉BF₁₂O₃
Molecular Weight: 403.958 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22124055
CAS-Nummer: 2317-76-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR